Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009288.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009288.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009288.hg.1 |
TC11000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009287.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009287.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009287.hg.1 |
TC11000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100160524.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100160524.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100160524.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100160524.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100160524.hg.1 |
TC1100008199.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000265643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
ENST00000538401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008199.hg.1 |
NM_015973.4 |
Refseq |