Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
ENST00000557681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014708.hg.1 |
ENST00000334696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014708.hg.1 |
ENST00000557325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014708.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014708.hg.1 |
TC14001302.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171174.hg.1 |
TC1400009682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000334696.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000553284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000554664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000555829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556108.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000556242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
ENST00000557681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009682.hg.1 |
NM_001249.3 |
Refseq |