Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000459.4 | NP_000450.2 | 20 - 21 | 3556 | 92 | |
NM_001290077.1 | NP_001277006.1 | 19 - 20 | 3427 | 92 | ||
NM_001290078.1 | NP_001277007.1 | 18 - 19 | 3112 | 92 | ||
XM_005251561.1 | XP_005251618.1 | 20 - 21 | 3220 | 92 | ||
XM_005251563.1 | XP_005251620.1 | 19 - 20 | 3091 | 92 | ||
GenBank mRNA | AB208796.1 | - | 20 - 21 | 3234 | 92 | |
AK291775.1 | - | 20 - 21 | 3244 | 92 | ||
AK294887.1 | - | 18 - 19 | 3113 | 92 | ||
AK295043.1 | - | 19 - 20 | 3153 | 92 | ||
BC035514.1 | - | 20 - 21 | 3231 | 5228999 | 92 | |
L06139.1 | - | 20 - 21 | 3254 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons |