Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: DNAH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206927.1 | NP_001193856.1 | 19 - 20 | 2780 | 87 | |
XM_011514318.2 | XP_011512620.1 | 19 - 20 | 2791 | 87 | ||
XM_011514319.2 | XP_011512621.1 | 19 - 20 | 2791 | 87 | ||
XM_011514320.2 | XP_011512622.1 | 17 - 18 | 2554 | 87 | ||
XM_011514321.1 | XP_011512623.1 | 16 - 17 | 2138 | 87 | ||
XM_017010325.1 | XP_016865814.1 | 19 - 20 | 2791 | 87 | ||
XM_017010326.1 | XP_016865815.1 | 19 - 20 | 2791 | 87 | ||
XM_017010327.1 | XP_016865816.1 | 19 - 20 | 2792 | 87 | ||
GenBank mRNA | AF356519.1 | - | 17 - 18 | 2274 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |