Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000278175.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000524948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000525063.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000526492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000528655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000530439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
ENST00000534464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006831.hg.1 |
NM_001124.2 |
Refseq |