Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022569.hg.1 |
ENST00000316878.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022569.hg.1 |
ENST00000501501.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022569.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022569.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022569.hg.1 |
TC17001679.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022568.hg.1 |
ENST00000316878.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022568.hg.1 |
ENST00000501501.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022568.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022568.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022568.hg.1 |
TC17001679.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011084.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189197.hg.1 |
TC1700011084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
ENST00000513579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
ENST00000612501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
NM_032595.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189196.hg.1 |
TC1700011084.hg.1 |
Clariom D TC |