Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002825.hg.1 |
TC6_qbl_hap6000152.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002825.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002373.hg.1 |
TC6_mann_hap4000137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002373.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002843.hg.1 |
TC6_cox_hap2000160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002843.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002757.hg.1 |
TC6_dbb_hap3000149.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002757.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017516.hg.1 |
TC06001531.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017516.hg.1 |
ENST00000376073.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017516.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017516.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001162.hg.1 |
TC6_apd_hap1000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001162.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002514.hg.1 |
TC6_mcf_hap5000137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002514.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002513.hg.1 |
TC6_ssto_hap7000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002513.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177294.hg.1 |
TC0600011439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000340027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000376073.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000491161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
ENST00000495600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_001145467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011439.hg.1 |
NM_147130.2 |
Refseq |