Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038046.hg.1 |
TC02002424.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038046.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038046.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038046.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011463.hg.1 |
ENST00000476891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038044.hg.1 |
TC02002424.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038044.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038044.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038048.hg.1 |
TC02002424.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038048.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038048.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011463.hg.1 |
ENST00000476891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206074.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206074.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206074.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206074.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206074.hg.1 |
TC0200014609.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206077.hg.1 |
TC0200014609.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
ENST00000409092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014609.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206079.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206079.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206079.hg.1 |
ENST00000331426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206079.hg.1 |
NM_198557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206079.hg.1 |
TC0200014609.hg.1 |
Clariom D TC |