Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104629.1 | NP_001098099.1 | 2 - 3 | 160 | 138 | |
GenBank mRNA | BC037906.1 | - | 2 - 3 | 121 | 5276648 | 138 |
CF995148.1 | - | 2 - 3 | 164 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |