Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030849.hg.1 |
TC02001945.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030849.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030849.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200194087.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200194087.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200194087.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200194087.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200194087.hg.1 |
TC0200012946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012946.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200194088.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200194088.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200194088.hg.1 |
ENST00000295379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200194088.hg.1 |
NM_014482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200194088.hg.1 |
TC0200012946.hg.1 |
Clariom D TC |