Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ariadne homolog 2 opposite strand | ARIH2OS | C3orf71 | -17.1kb | Anti-sense | Chr. 3 - 48917788 - 48919385 on NCBI Build GRCh38 |
Downstream |
prolyl 4-hydroxylase, transmembrane |
P4HTM |
EGLN4, HIFPH4, P4H-TM, PH-4, PH4, PHD4 |
50.2kb | Sense | Chr. 3 - 48989908 - 49007149 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |