Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: RPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283419.1 | NP_001270348.1 | 4 | 1220 | 126 | |
XM_015451028.1 | XP_015306514.1 | 4 | 1295 | 126 | ||
GenBank mRNA | AB169560.1 | - | 4 | 1225 | 126 | |
AB170836.1 | - | 4 | 1220 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |