Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025899.2 | NP_080175.1 | 8 - 9 | 776 | 63 | |
GenBank mRNA | AK004492.1 | - | 8 - 9 | 751 | 63 | |
AK005151.1 | - | 8 - 9 | 760 | 63 | ||
AK007669.1 | - | 7 - 8 | 640 | 63 | ||
AK075856.1 | - | 8 - 9 | 720 | 63 | ||
AK077583.1 | - | 8 - 9 | 725 | 63 | ||
AK088103.1 | - | 8 - 9 | 730 | 63 | ||
AK167377.1 | - | 8 - 9 | 730 | 63 | ||
BC003423.1 | - | 8 - 9 | 748 | 2609295 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |