Search Thermo Fisher Scientific
Gene Symbol: Nudt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290182.1 | NP_001277111.1 | 1 - 2 | 130 | 69 | |
NM_024437.4 | NP_077757.2 | 1 - 2 | 130 | 69 | ||
NM_024446.5 | NP_077766.3 | 1 - 2 | 130 | 69 | ||
GenBank mRNA | AF338424.1 | - | 1 - 2 | 90 | 69 | |
AK008824.1 | - | 1 - 2 | 57 | 69 | ||
AK011172.1 | - | 1 - 2 | 64 | 69 | ||
BC033046.1 | - | 1 - 2 | 62 | 69 | ||
BC069843.1 | - | 1 - 2 | 86 | 5102473 | 69 | |
CA460240.1 | - | 1 - 2 | 162 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |