Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289709.1 | NP_001276638.1 | 4 - 5 | 578 | 78 | |
NM_001289710.1 | NP_001276639.1 | 4 - 5 | 578 | 78 | ||
NM_139302.2 | NP_647463.1 | 4 - 5 | 578 | 78 | ||
GenBank mRNA | AF426314.1 | - | 4 - 5 | 475 | 78 | |
AK129455.1 | - | 4 - 5 | 506 | 78 | ||
AK152409.1 | - | 4 - 5 | 508 | 78 | ||
AK152502.1 | - | 4 - 5 | 529 | 78 | ||
AK155248.1 | - | 4 - 5 | 503 | 78 | ||
AK167762.1 | - | 4 - 5 | 551 | 78 | ||
BC024477.1 | - | 4 - 5 | 493 | 4977001 | 78 | |
BC028859.1 | - | 4 - 5 | 567 | 4167974 | 78 | |
EF676083.1 | - | 4 - 5 | 554 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |