Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 23 - 24 | 9384 | 80 | |
NM_009765.3 | NP_033895.2 | 23 - 24 | 9190 | 80 | ||
GenBank mRNA | BC059904.1 | - | 13 - 14 | 2647 | 80 | |
U65594.1 | - | 23 - 24 | 9167 | 80 | ||
U82270.1 | - | 22 - 23 | 8876 | 80 | ||
U89652.1 | - | 22 - 23 | 8914 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |