Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042527.2 | NP_001035992.1 | 9 - 10 | 2293 | 132 | |
NM_007550.4 | NP_031576.4 | 9 - 10 | 2564 | 132 | ||
GenBank mRNA | AB008674.1 | - | 9 - 10 | 2554 | 132 | |
AK137022.1 | - | 9 - 10 | 2276 | 132 | ||
AK157898.1 | - | 9 - 10 | 2294 | 132 | ||
AK164605.1 | - | 9 - 10 | 2260 | 132 | ||
Z98263.1 | - | 9 - 10 | 2271 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |