Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011936.2 | NP_036066.2 | 4 - 5 | 938 | 75 | |
GenBank mRNA | AJ237917.1 | - | 4 - 5 | 924 | 75 | |
AK040866.1 | - | 4 - 5 | 916 | 75 | ||
AK045465.1 | - | 4 - 5 | 915 | 75 | ||
AK046935.1 | - | 4 - 5 | 897 | 75 | ||
AK049502.1 | - | 4 - 5 | 931 | 75 | ||
AK088881.1 | - | 4 - 5 | 924 | 75 | ||
AK129437.1 | - | 4 - 5 | 905 | 75 | ||
AK161060.1 | - | 4 - 5 | 897 | 75 | ||
BC022222.1 | - | 4 - 5 | 889 | 4237261 | 75 | |
BC057008.1 | - | 4 - 5 | 904 | 5708558 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |