Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Copz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019817.1 | NP_062791.1 | 3 - 4 | 176 | 147 | |
GenBank mRNA | AB037370.1 | - | 3 - 4 | 176 | 147 | |
AK003302.1 | - | 3 - 4 | 184 | 147 | ||
AK083824.1 | - | 3 - 4 | 196 | 147 | ||
AK156758.1 | - | 3 - 4 | 195 | 147 | ||
AK167636.1 | - | 3 - 4 | 184 | 147 | ||
AK168382.1 | - | 3 - 4 | 195 | 147 | ||
AK168471.1 | - | 3 - 4 | 195 | 147 | ||
BC002246.1 | - | 3 - 4 | 182 | 147 | ||
BC023207.1 | - | 2 - 3 | 160 | 147 | ||
BC025041.1 | - | 3 - 4 | 182 | 4954031 | 147 | |
BC039920.1 | - | 3 - 4 | 205 | 147 | ||
BC052891.1 | - | 3 - 4 | 187 | 147 | ||
BC058524.1 | - | 3 - 4 | 175 | 5691652 | 147 | |
BC062203.1 | - | 3 - 4 | 192 | 147 | ||
BC085314.1 | - | 3 - 4 | 197 | 30441470 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |