Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033654.2 | NP_001028826.1 | 2 - 3 | 200 | 107 | |
NM_028304.2 | NP_082580.1 | 2 - 3 | 613 | 107 | ||
NM_028956.4 | NP_083232.1 | 2 - 3 | 210 | 107 | ||
GenBank mRNA | AK012737.1 | - | 2 - 3 | 377 | 107 | |
AK017063.1 | - | 2 - 3 | 187 | 107 | ||
AK028368.1 | - | 2 - 3 | 197 | 107 | ||
AK029866.1 | - | 2 - 3 | 201 | 107 | ||
AK087766.1 | - | 2 - 3 | 392 | 107 | ||
AK088562.1 | - | 2 - 3 | 201 | 107 | ||
BC025555.1 | - | 2 - 3 | 431 | 5321811 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |