Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Slc6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146013.1 | NP_001139485.1 | 7 - 8 | 1337 | 65 | |
NM_148931.3 | NP_683733.2 | 6 - 7 | 1255 | 65 | ||
GenBank mRNA | AF411042.3 | - | 7 - 8 | 1391 | 65 | |
AY147186.2 | - | 7 - 8 | 1490 | 65 | ||
BC138125.1 | - | 6 - 7 | 1258 | 8861144 | 65 | |
BC138126.1 | - | 6 - 7 | 1258 | 8861145 | 65 | |
BC157997.1 | - | 7 - 8 | 1337 | 9007308 | 65 | |
BC172125.1 | - | 6 - 7 | 1198 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |