Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174866.2 | NP_777355.1 | 2 - 3 | 212 | 75 | |
GenBank mRNA | BC044756.2 | - | 1 - 2 | 187 | 75 | |
BC128019.1 | - | 2 - 3 | 215 | 40136266 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |