Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011772.2 | NP_035902.2 | 4 - 5 | 730 | 75 | |
GenBank mRNA | AB017615.1 | - | 3 - 4 | 1137 | 75 | |
AK089522.1 | - | 4 - 5 | 564 | 75 | ||
AK165740.1 | - | 994 | 75 | |||
AK170453.1 | - | 5 - 6 | 654 | 75 | ||
BC141286.1 | - | 3 - 4 | 828 | 9055898 | 75 | |
BC145614.1 | - | 3 - 4 | 814 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |