Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138955.3 | NP_620405.3 | 13 - 14 | 1838 | 51 | |
GenBank mRNA | AF378330.1 | - | 12 - 13 | 1669 | 51 | |
AF411084.1 | - | 13 - 14 | 1716 | 51 | ||
AF425077.1 | - | 13 - 14 | 1718 | 51 | ||
AJ426047.2 | - | 13 - 14 | 1717 | 51 | ||
AK043147.1 | - | 13 - 14 | 1839 | 51 | ||
AK157317.1 | - | 13 - 14 | 2163 | 51 | ||
AY040865.1 | - | 13 - 14 | 1682 | 51 | ||
BC016200.1 | - | 13 - 14 | 1718 | 4511456 | 51 | |
BC023077.1 | - | 7 - 8 | 926 | 51 | ||
BC059033.1 | - | 4 - 5 | 1011 | 51 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |