Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176954.3 | NP_795928.2 | 9 - 10 | 1097 | 124 | |
GenBank mRNA | AK029796.1 | - | 9 - 10 | 2271 | 124 | |
AK132321.1 | - | 9 - 10 | 1063 | 124 | ||
AK158669.1 | - | 8 - 9 | 874 | 124 | ||
BC079845.1 | - | 9 - 10 | 1068 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |