Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010823.3 | NP_034953.2 | 4 - 5 | 706 | 75 | |
GenBank mRNA | AK041166.1 | - | 4 - 5 | 678 | 75 | |
BC103513.1 | - | 4 - 5 | 663 | 75 | ||
BC146612.1 | - | 4 - 5 | 659 | 75 | ||
BC146613.1 | - | 4 - 5 | 664 | 40039147 | 75 | |
Z22649.1 | - | 4 - 5 | 664 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |