Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146013.1 | NP_001139485.1 | 11 - 12 | 70 | ||
NM_148931.3 | NP_683733.2 | 10 - 11 | 70 | |||
GenBank mRNA | AB118159.1 | - | 11 - 12 | 1771 | 70 | |
AF411042.3 | - | 11 - 12 | 1878 | 70 | ||
AY147186.2 | - | 11 - 12 | 1977 | 70 | ||
BC138125.1 | - | 10 - 11 | 8861144 | 70 | ||
BC138126.1 | - | 10 - 11 | 8861145 | 70 | ||
BC157997.1 | - | 11 - 12 | 9007308 | 70 | ||
BC172125.1 | - | 10 - 11 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |