Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133740.2 | NP_598501.1 | 2 - 3 | 272 | 108 | |
GenBank mRNA | AK008118.1 | - | 2 - 3 | 264 | 108 | |
AK010534.1 | - | 2 - 3 | 284 | 108 | ||
AK031738.1 | - | 2 - 3 | 273 | 108 | ||
AK049836.1 | - | 2 - 3 | 253 | 108 | ||
AK086646.1 | - | 2 - 3 | 231 | 108 | ||
AK155236.1 | - | 2 - 3 | 270 | 108 | ||
BC050775.1 | - | 2 - 3 | 251 | 6534023 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |