Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027376.3 | NP_081652.2 | 6 - 7 | 862 | 71 | |
GenBank mRNA | AK033247.1 | - | 6 - 7 | 849 | 71 | |
AK045745.1 | - | 857 | 71 | |||
AK134650.1 | - | 6 - 7 | 862 | 71 | ||
AK168939.1 | - | 6 - 7 | 844 | 71 | ||
BC045601.1 | - | 5 - 6 | 1726 | 71 | ||
CK624547.1 | - | 1 - 2 | 124 | 71 | ||
CO427180.1 | - | 4 - 5 | 435 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |