Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009796.2 | NP_033926.1 | 19 - 20 | 2360 | 137 | |
GenBank mRNA | AB028640.1 | - | 19 - 20 | 2360 | 137 | |
AJ012475.1 | - | 19 - 20 | 2173 | 137 | ||
AK154277.1 | - | 19 - 20 | 2446 | 137 | ||
AK169334.1 | - | 19 - 20 | 3405 | 137 | ||
BC095980.1 | - | 19 - 20 | 3375 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |