Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222421_m1
          See other KDR GE Assays ›
          Gene Symbol:
          Kdr
          Gene Name:
          kinase insert domain protein receptor
          Gene Aliases:
          6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2
          Chromosome Location:
          Chr.5: 75932827 - 75979074 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222421_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010612.2 NP_034742.2 13 - 14 2246 64
          GenBank mRNA BC020530.1 - 13 - 14 2246 4238984 64
          S53103.1 - 13 - 14 2257 64
          X59397.1 - 13 - 14 2193 64
          X70842.1 - 13 - 14 2271 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002623.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kdr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovarian follicle development
          vasculogenesis
          response to hypoxia
          positive regulation of protein phosphorylation
          regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lymph vessel development
          cell migration involved in sprouting angiogenesis
          positive regulation of mesenchymal cell proliferation
          endocardium development
          endothelium development
          endochondral bone growth
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cytosolic calcium ion concentration
          multicellular organism development
          positive regulation of cell proliferation
          regulation of cell shape
          male gonad development
          positive regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          cell differentiation
          lung development
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of TOR signaling
          embryonic hemopoiesis
          calcium-mediated signaling using intracellular calcium source
          cellular response to vascular endothelial growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          vascular endothelial growth factor signaling pathway
          response to drug
          negative regulation of apoptotic process
          surfactant homeostasis
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          cell fate commitment
          endothelial cell differentiation
          regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of vasodilation
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          positive regulation of long-term neuronal synaptic plasticity
          lung alveolus development
          cell maturation
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          positive regulation of epithelial cell proliferation
          positive regulation of calcium-mediated signaling
          positive regulation of positive chemotaxis
          positive regulation of nitric-oxide synthase biosynthetic process
          positive regulation of focal adhesion assembly
          calcium ion homeostasis
          blood vessel endothelial cell differentiation
          positive regulation of ERK1 and ERK2 cascade
          regulation of hematopoietic progenitor cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of vasculogenesis
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          vascular endothelial growth factor-activated receptor activity
          integrin binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          growth factor binding
          vascular endothelial growth factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500023207.mm.1 NM_010612.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023207.mm.1 NM_010612.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023207.mm.1 TC0500002623.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-kzhm8:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline