Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030033.1 | NP_084309.1 | 1 - 2 | 67 | 87 | |
GenBank mRNA | AK020340.1 | - | 2 - 3 | 128 | 87 | |
AY705449.1 | - | 1 - 2 | 67 | 87 | ||
DQ200908.1 | - | 3 - 4 | 244 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |