Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 1 - 2 | 491 | 110 | |
GenBank mRNA | AK166167.1 | - | 1 - 2 | 466 | 110 | |
AK172567.1 | - | 1 - 2 | 457 | 110 | ||
AK173313.1 | - | 1 - 2 | 110 | 110 | ||
BC010833.1 | - | 1 - 2 | 106 | 4235165 | 110 | |
BC054121.1 | - | 1 - 2 | 477 | 6393910 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |