Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146032.3 | NP_666144.3 | 1 - 2 | 297 | 78 | |
GenBank mRNA | AK033010.1 | - | 1 - 2 | 216 | 78 | |
AK076003.1 | - | 1 - 2 | 194 | 78 | ||
AK180890.1 | - | 1 - 2 | 52 | 78 | ||
AK212180.1 | - | 1 - 2 | 91 | 78 | ||
AK212446.1 | - | 1 - 2 | 91 | 78 | ||
AK213152.1 | - | 1 - 2 | 91 | 78 | ||
BC070422.1 | - | 1 - 2 | 297 | 6403747 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |