Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128080.2 | NP_001121552.1 | 1 - 2 | 128 | 82 | |
GenBank mRNA | AK041748.1 | - | 1 - 2 | 114 | 82 | |
AY039000.1 | - | 1 - 2 | 136 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |