Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Postn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198765.1 | NP_001185694.1 | 14 - 15 | 1934 | 131 | |
NM_001198766.1 | NP_001185695.1 | 14 - 15 | 1934 | 131 | ||
NM_015784.3 | NP_056599.1 | 14 - 15 | 1934 | 131 | ||
GenBank mRNA | AK030756.1 | - | 14 - 15 | 1933 | 131 | |
BC007141.1 | - | 14 - 15 | 1928 | 131 | ||
BC031449.1 | - | 14 - 15 | 1903 | 4457222 | 131 | |
D13664.1 | - | 14 - 15 | 1916 | 131 | ||
E05655.1 | - | 14 - 15 | 1930 | 131 | ||
KM117171.1 | - | 14 - 15 | 1898 | 131 | ||
KM117172.1 | - | 14 - 15 | 1898 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |