Search Thermo Fisher Scientific
Gene Symbol: Alg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025442.3 | NP_079718.1 | 3 - 4 | 326 | 82 | |
GenBank mRNA | AK003818.1 | - | 3 - 4 | 307 | 82 | |
AK005296.1 | - | 3 - 4 | 327 | 82 | ||
AK030567.1 | - | 3 - 4 | 297 | 82 | ||
AK161187.1 | - | 305 | 82 | |||
BC027160.1 | - | 3 - 4 | 286 | 4946430 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |