Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177461.1 | NP_803412.1 | 14 - 15 | 2743 | 105 | |
GenBank mRNA | AK007847.1 | - | 1 - 2 | 172 | 105 | |
AK076226.1 | - | 1 - 2 | 44 | 105 | ||
AK076763.1 | - | 8 - 9 | 1400 | 105 | ||
AK077217.1 | - | 7 - 8 | 1571 | 105 | ||
AK129419.1 | - | 14 - 15 | 2481 | 105 | ||
AK147587.1 | - | 14 - 15 | 2544 | 105 | ||
AK147646.1 | - | 14 - 15 | 2756 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |