Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080158.1 | NP_001073627.1 | 1 - 2 | 201 | 70 | |
NM_025639.4 | NP_079915.1 | 2 - 3 | 202 | 70 | ||
NM_178269.3 | NP_840000.2 | 2 - 3 | 202 | 70 | ||
GenBank mRNA | AK010885.1 | - | 2 - 3 | 194 | 70 | |
AK011191.1 | - | 2 - 3 | 179 | 70 | ||
AK011227.1 | - | 2 - 3 | 186 | 70 | ||
AK011464.1 | - | 2 - 3 | 203 | 70 | ||
AK011755.1 | - | 2 - 3 | 195 | 70 | ||
AK011793.1 | - | 2 - 3 | 188 | 70 | ||
AK017679.1 | - | 2 - 3 | 195 | 70 | ||
AK186157.1 | - | 1 - 2 | 40 | 70 | ||
AY243536.1 | - | 2 - 3 | 163 | 70 | ||
AY243537.1 | - | 2 - 3 | 163 | 70 | ||
BC009160.1 | - | 2 - 3 | 181 | 2655787 | 70 | |
BG276334.1 | - | 1 - 2 | 201 | 70 | ||
BY710734.1 | - | 2 - 3 | 203 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |