Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026083.2 | NP_080359.2 | 3 - 4 | 532 | 91 | |
GenBank mRNA | AK014200.1 | - | 3 - 4 | 312 | 91 | |
AK078225.1 | - | 3 - 4 | 523 | 91 | ||
BB619434.1 | - | 3 - 4 | 533 | 91 | ||
BC138264.1 | - | 3 - 4 | 481 | 8861283 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |