Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029453.2 | NP_083729.2 | 16 - 17 | 2274 | 99 | |
GenBank mRNA | AK015153.1 | - | 4 - 5 | 507 | 99 | |
AK015753.1 | - | 5 - 6 | 727 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |