Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081057.2 | NP_001074526.1 | 15 - 16 | 3534 | 105 | |
NM_001289510.1 | NP_001276439.1 | 14 - 15 | 3482 | 105 | ||
NM_175336.3 | NP_780545.2 | 15 - 16 | 3644 | 105 | ||
GenBank mRNA | AK129113.1 | - | 14 - 15 | 3486 | 105 | |
AK147592.1 | - | 14 - 15 | 3482 | 105 | ||
BC041778.1 | - | 12 - 13 | 3126 | 105 | ||
BC138354.1 | - | 14 - 15 | 3477 | 8861374 | 105 | |
BC145250.1 | - | 14 - 15 | 3587 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |