Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem30c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027651.1 | NP_081927.1 | 4 - 5 | 483 | 148 | |
GenBank mRNA | AK016597.1 | - | 4 - 5 | 497 | 148 | |
AK161475.1 | - | 3 - 4 | 2347 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Probe spans exons |