Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290792.1 | NP_001277721.1 | 1 - 2 | 153 | 65 | |
NM_001290795.1 | NP_001277724.1 | 1 - 2 | 153 | 65 | ||
GenBank mRNA | AF229635.1 | - | 1 - 2 | 127 | 65 | |
AK018409.1 | - | 1 - 2 | 149 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |