| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10603952 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603976 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599097 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603979 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599078 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603957 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603972 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603924 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603971 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603989 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603959 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599081 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599076 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603950 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599065 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604015 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603990 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599101 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603934 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603960 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603931 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604016 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599100 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604011 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603930 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603993 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603982 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603991 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603980 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603921 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603917 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599080 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603978 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603935 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599104 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599099 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603922 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603951 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603958 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599077 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604009 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604013 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603987 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603994 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603933 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603927 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603923 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603981 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604012 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604010 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603949 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599103 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599102 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603988 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603916 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599098 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603925 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603961 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603992 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10604014 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603977 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599079 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10603915 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10603914 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10603926 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0X00002017.mm.1 |
NM_001110250.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002017.mm.1 |
NM_009529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002017.mm.1 |
TC0X00000308.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012548.mm.1 |
NM_001025607.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012548.mm.1 |
NM_001025607.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012548.mm.1 |
TC0X00001978.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012601.mm.1 |
NM_001162364.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012601.mm.1 |
TC0X00001985.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012700.mm.1 |
NM_001100610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012700.mm.1 |
NM_001109969.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012700.mm.1 |
NM_001109969.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012700.mm.1 |
TC0X00001995.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012474.mm.1 |
NM_001099919.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012474.mm.1 |
NM_001102678.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012474.mm.1 |
TC0X00001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012495.mm.1 |
NM_001040669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012495.mm.1 |
NM_001040669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012495.mm.1 |
TC0X00001971.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012589.mm.1 |
NM_001136476.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012589.mm.1 |
NM_001136476.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012589.mm.1 |
TC0X00001983.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012590.mm.1 |
NM_001136476.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012590.mm.1 |
NM_001136476.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012590.mm.1 |
TC0X00001983.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012559.mm.1 |
NM_001104946.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012559.mm.1 |
NM_001104946.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012559.mm.1 |
TC0X00001979.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012769.mm.1 |
NM_001099325.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012769.mm.1 |
NM_001100609.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012769.mm.1 |
TC0X00002002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00001871.mm.1 |
NM_001099325.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001871.mm.1 |
NM_001109970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001871.mm.1 |
NM_001110250.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001871.mm.1 |
TC0X00000267.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00001872.mm.1 |
NM_001099325.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001872.mm.1 |
NM_001109970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001872.mm.1 |
NM_001110250.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00001872.mm.1 |
TC0X00000267.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002016.mm.1 |
NM_001110250.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002016.mm.1 |
NM_009529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002016.mm.1 |
TC0X00000308.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012435.mm.1 |
NM_001100444.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012435.mm.1 |
TC0X00001964.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012457.mm.1 |
NM_001081657.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012457.mm.1 |
NM_001081657.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012457.mm.1 |
TC0X00001968.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012496.mm.1 |
NM_001040669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012496.mm.1 |
NM_001040669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012496.mm.1 |
TC0X00001971.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012701.mm.1 |
NM_001100610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012701.mm.1 |
NM_001109969.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012701.mm.1 |
NM_001109969.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012701.mm.1 |
TC0X00001995.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012434.mm.1 |
NM_001100444.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012434.mm.1 |
TC0X00001964.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012437.mm.1 |
NM_001100444.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012437.mm.1 |
TC0X00001964.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012458.mm.1 |
NM_001081657.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012458.mm.1 |
NM_001081657.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012458.mm.1 |
TC0X00001968.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012547.mm.1 |
NM_001025607.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012547.mm.1 |
NM_001025607.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012547.mm.1 |
TC0X00001978.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012602.mm.1 |
NM_001162364.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012602.mm.1 |
TC0X00001985.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012651.mm.1 |
NM_001099347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012651.mm.1 |
NM_001099347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012651.mm.1 |
TC0X00001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012436.mm.1 |
NM_001100444.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012436.mm.1 |
TC0X00001964.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012477.mm.1 |
NM_001099919.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012477.mm.1 |
NM_001102678.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012477.mm.1 |
TC0X00001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012560.mm.1 |
NM_001104946.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012560.mm.1 |
NM_001104946.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012560.mm.1 |
TC0X00001979.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012619.mm.1 |
NM_001114754.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012619.mm.1 |
NM_001114754.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012619.mm.1 |
TC0X00001987.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012620.mm.1 |
NM_001114754.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012620.mm.1 |
NM_001114754.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012620.mm.1 |
TC0X00001987.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012770.mm.1 |
NM_001099325.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012770.mm.1 |
NM_001100609.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012770.mm.1 |
TC0X00002002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012652.mm.1 |
NM_001099347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012652.mm.1 |
NM_001099347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012652.mm.1 |
TC0X00001990.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1422618_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1422617_at |
- |
- |
| Mouse Expression Set 430 |
1422618_x_at |
- |
- |
| Mouse Expression Set 430 |
1422617_PM_at |
- |
- |
| Murine Genome U74v2 Set |
102818_PM_at |
- |
- |
| Murine Genome U74v2 Set |
102818_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1422618_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1422617_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1422618_PM_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1422617_PM_at |
- |
- |