Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00004253.mm.1 |
NM_001111293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004253.mm.1 |
NM_001111293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004253.mm.1 |
TC0X00000675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014948.mm.1 |
NM_031493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014948.mm.1 |
NM_031493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014948.mm.1 |
TC0X00003428.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014935.mm.1 |
NM_001045539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014935.mm.1 |
NM_001045539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014935.mm.1 |
TC0X00003427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004252.mm.1 |
NM_001111293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004252.mm.1 |
NM_001111293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004252.mm.1 |
TC0X00000675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004250.mm.1 |
NM_001111293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004250.mm.1 |
NM_001111293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004250.mm.1 |
TC0X00000675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014947.mm.1 |
NM_031493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014947.mm.1 |
NM_031493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014947.mm.1 |
TC0X00003428.mm.1 |
MTA TC |