Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308589.1 | NP_001295518.1 | 3 - 4 | 807 | 92 | |
GenBank mRNA | KF873613.1 | - | 3 - 4 | 807 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |