Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC101117682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012902.4 | XP_004012951.2 | 4 - 5 | 898 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |