Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102069.3 | XP_001102069.1 | 931 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |