Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ING2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193975.2 | NP_001180904.1 | 375 | 114 | ||
GenBank mRNA | DQ147943.1 | - | 145 | 114 | ||
JU320774.1 | - | 187 | 114 | |||
JU472479.1 | - | 308 | 114 | |||
JV045131.1 | - | 308 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |